missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caldesmon/CALD1 Antibody (rCALD1/820), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08326JF549
This item is not returnable.
View return policy
Description
Caldesmon/CALD1 Monoclonal antibody specifically detects Caldesmon/CALD1 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Specifications
| Caldesmon/CALD1 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| CAD, caldesmon, caldesmon 1, CDMH-CAD, HCAD, LCAD, L-CAD, MGC21352, NAG22 | |
| Recombinant full-length human Caldesmon/CALD1 protein (Uniprot: Q05682 ) | |
| 100 μg | |
| Cell Biology, Cellular Markers | |
| 800 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Immunohistochemistry (Paraffin) | |
| rCALD1/820 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction