Learn More
Invitrogen™ Caldesmon Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5117825
Product Code. 17491483
  • € 476.00 / 100µL
Description
Caldesmon Polyclonal Antibody for Western Blot, ICC/IF, IP
Caldesmon is a developmentally regulated protein involved in smooth muscle and non muscle contraction. Two closely related variants of human caldesmon have been identified which differ in their electrophorectic mobility and cellular distribution. The h caldesmon variant (120 - 150 kDa) is predominantly expressed in smooth muscle whereas l caldesmon (70 - 80 kDa) is found in non-muscle tissue and cells. Neither of the two variants has been detected in skeletal muscle.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Caldesmon
Polyclonal
PBS with 0.03% ProClin 300; pH 7
Q05682
Cald1
E. coli-derived Human CALD1/caldesmon fragment.
100 μL
Primary
Human
Antibody
IgG
Immunoprecipitation, Western Blot, Immunocytochemistry
Unconjugated
Cald1
4833423D12Rik; AI195384; AV071549; AW536160; C920027I18Rik; CAD; Cald1; Caldesmon; caldesmon 1; CDM; HCAD; H-CAD; h-caldesmon; LCAD; L-CAD; L-caldesmon; MGC21352; NAG22; Non-muscle caldesmon; OTTHUMP00000207950; OTTHUMP00000207953; OTTHUMP00000207954; OTTHUMP00000207955; OTTHUMP00000214473; smooth muscle caldesmon; testis secretory sperm-binding protein Li 227n
Rabbit
Antigen affinity chromatography, Protein A
RUO
800
-20°C, Avoid Freeze/Thaw Cycles