Learn More
Caldesmon Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 20887-1-AP-20UL
 View more versions of this product
Product Code. 16890465
  • € 145.00 / 20µL
Description
Caldesmon is a developmentally regulated protein involved in smooth muscle and non-muscle contraction. Two closely related variants of human caldesmon have been identified which differ in their eletrophoretic mobility and cellular distribution. The h-caldesmon variant (120-150kDa) is predomitly expressed in smooth muscle whereas l-caldesmon (70-80kDa) is found in non-muscle tissue and cells. Neither of the two variants has been detected in skeletal muscle.
Specifications
Specifications
Caldesmon
Polyclonal
Unconjugated
CALD1
CAD, CALD1, Caldesmon, caldesmon 1, CDM, H CAD, L CAD, NAG22
Rabbit
Antigen Affinity Chromatography
RUO
109624, 800
-20°C
Liquid
Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q05682
CALD1
Caldesmon Fusion Protein Ag13676
20 μL
Primary
Human, Mouse