Learn More
Caldesmon Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20887-1-AP-20UL
This item is not returnable.
View return policy
Description
Caldesmon is a developmentally regulated protein involved in smooth muscle and non-muscle contraction. Two closely related variants of human caldesmon have been identified which differ in their eletrophoretic mobility and cellular distribution. The h-caldesmon variant (120-150kDa) is predomitly expressed in smooth muscle whereas l-caldesmon (70-80kDa) is found in non-muscle tissue and cells. Neither of the two variants has been detected in skeletal muscle.Specifications
| Caldesmon | |
| Polyclonal | |
| Unconjugated | |
| CALD1 | |
| CAD, CALD1, Caldesmon, caldesmon 1, CDM, H CAD, L CAD, NAG22 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 109624, 800 | |
| -20°C | |
| Liquid |
| Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q05682 | |
| CALD1 | |
| Caldesmon Fusion Protein Ag13676 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |