missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CALHM3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17180914 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17180914 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17180914 Supplier Invitrogen™ Supplier No. PA5113057

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CALHM3 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

CALHM3 is a pore-forming subunit of a voltage-gated ion channel.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CALHM3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene CALHM3
Gene Accession No. Q86XJ0
Gene Alias bA225H22.7; calcium homeostasis modulator 3; calcium homeostasis modulator protein 3; Calhm3; FAM26A; family with sequence similarity 26, member A; Protein FAM26A; RGD1560215
Gene Symbols CALHM3
Host Species Rabbit
Immunogen Recombinant Human Calcium homeostasis modulator protein 3 protein (201-350AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 119395, 240669
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.