Learn More
Endecotts™ Sieve Calibration Standards
Description
- Calibrates sieves to an accuracy of ±1μm
- Soda-lime glass microspheres pass over the entire surface of the sieve and through every aperture.
- Traceable to the United Kingdom National Physical Laboratory
- Standards include a calibration chart.
Specifications
Specifications
| Product Type | Calibration Ball Mesh |
| Material | Soda-lime glass |
| For Use With (Equipment) | Sieve |
| Packaging Quantity | 5/Pk. |
| Particle Size | 212 μm |