missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calibration Buffer Solutions, Hanna Instruments™
Click to view available options
Quantity:
20mL
1L
pH:
4.01
9.18
10.01
Description
- A premium quality pH 4.01 calibration solution that has been dyed red for easy buffer identification
- pH 4.01 calibration buffer produced in accordance with ISO 3696/BS3978 standards
- Uses high purity salts, deionized water, certified weight-checked balances, and Class A glassware in a temperature controlled environment monitored with certified thermometers
- Reported values are accurate to +/- 0.01 pH @ 25°C and are traceable to NIST Standard Reference Materials (SRMs)
Specifications
Color | Colorless |
Composition | Aqueous Buffer Solution |
Physical Form | Liquid |
pH | 4.01 |
Quantity | 1L |
Solution Type | Buffer |
Odor | Odorless |