Learn More
Hanna Instruments™ Calibration Buffer Sachet
Click to view available options
pH Range:
4.01 & 7.01
Unit Size:
Each
Specifications
Specifications
| Accuracy (pH) | – 0.01 pH |
| Type | Calibration Buffer Sachet |
| For Use With (Application) | Ideal for users that do not calibrate frequently |
| pH Range | 4.01 & 7.01 |
| Certifications/Compliance | ISO 3696/BS3978 |