Learn More
BINDER™ Calibration Certificate, Temperature
Product Code. 15902962 Shop All BINDER Products
Click to view available options
For Use With (Equipment):
BINDER Chambers Series MK
BINDER Chambers Series MKF
BINDER Chambers Series MKFT
BINDER Chambers Series MKT
Calibration Points:
1 Measurement Point
15 to 18 Measurement Points
27 Measurement Points
9 Measurement Points
Unit Size:
Each
Product Code. 15902962
Brand: BINDER™ 80121143
Calibration Certificate for temperature, measurement in center of chamber at specified temperature.
  • Measurement at specified temperature
  • Including certificate
  • Various points of measurement
Specifications
For Use With (Equipment) BINDER Chambers Series MK
Description Calibration certificate, temperature - for temperature, measurement in center of chamber at specified temperature
Calibration Points 1 Measurement Point
Duration 60 to 120 min.
Product Type Calibration Certificate (Temperature)
For Use With (Application) Temperature Measurement