Learn More
BINDER™ Calibration Certificate, Temperature
Shop All BINDER Products
Click to view available options
For Use With (Equipment):
BINDER Chambers Series MK
BINDER Chambers Series MKF
BINDER Chambers Series MKFT
BINDER Chambers Series MKT
Calibration Points:
1 Measurement Point
15 to 18 Measurement Points
27 Measurement Points
9 Measurement Points
Unit Size:
Each
Description
- Measurement at specified temperature
- Including certificate
- Various points of measurement
Specifications
Specifications
| For Use With (Equipment) | BINDER Chambers Series MKT |
| Description | Calibration certificate, temperature - for temperature, measurement in center of chamber at specified temperature |
| Calibration Points | 1 Measurement Point |
| Duration | 60 to 120 min. |
| Product Type | Calibration Certificate (Temperature) |
| For Use With (Application) | Temperature Measurement |