Learn More
BINDER™ Calibration Certificate, Temperature and Humidity
Product Code. 15943012 Shop All BINDER Products
Click to view available options
For Use With (Equipment):
BINDER Chambers Series MKF
BINDER Chambers Series MKFT
Calibration Points:
1 Measuring Point
27 Measuring Points
Unit Size:
Each
Product Code. 15943012
Brand: BINDER™ 80121615
Calibration certificate, temperature and humidity - Measurement in center of chamber at 25°C / 60% RH or at specified test values
  • One humidity point of measurement
  • Including certificate
  • Humidity measurement point in center of chamber
  • Temperature (according to DIN12880) and humidity measurement at 25°C / 60% RH or at specified values
Specifications
For Use With (Equipment) BINDER Chambers Series MKF
Description Calibration certificate, temperature and humidity - temperature (according to DIN12880) and humidity measurement incl. certificate, 27 temperature measuring points and 1 humidity measuring point, at 25°C / 60 % RH or (at specified values)
Calibration Points 27 Measuring Points
Product Type Calibration Certificate (Temperature and Humidity)
For Use With (Application) Temperature and Humidity Measurement