Learn More
Xylem Calibration fluid - ISO17025 - (15 mL Dropper Bottle)
Click to view available options
Refractive Index:
1.33299
1.34026
1.34401
1.34782
1.34968
1.35015
1.35093
1.35171
1.36384
1.37233
1.38115
1.40978
1.42009
1.4308
1.44193
Conditionnement:
15mL
Description
- Calibration Fluid
- For Refractometers
Spécification
Spécification
| Type | Calibration Fluid |
| Refractive Index | 1.44193 |
| For Use With (Equipment) | Refractometer |