Learn More
Applied Biosystems™ Calibration Kit, 384-well
Product Code. 15840393 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 x 384 well plate
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15840393 1 x 384 well plate Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15840393 Supplier Applied Biosystems™ Supplier No. 4432476
  • ONLINE EXCLUSIVE PRICE
    € 3415.00
    € 2049.00 / Pack of 1
    Save € 1366.00
    40% Off
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
The 384-Well Calibration Kit is used to maintain your real-time PCR instrument with 384-well block
  • The 384-Well Calibration Kit is used to maintain your real-time PCR instrument with 384-well block.
  • The kit includes region of interest (ROI), background, and dye calibrations, as well as RNaseP verification.
  • Please see your instrument user guide for full instructions on use.
TRUSTED_SUSTAINABILITY
Specifications
Calibrated Dye Proprietary Mix
For Use With (Equipment) ViiA™ 7 Dx System, QuantStudio™ Dx
Product Type Calibration Kit
For Use With (Application) Real Time PCR (qPCR)
Quantity 1 x 384 well plate
Calibration Type Spectral Calibration
PCR Method qPCR