Learn More
Xylem Calibration Oil 5 x 5 mL Bottles (UKAS/NIST)
Product Code. p-200113487
Click to view available options
Refractive Index:
1.46666
1.55768
Unit Size:
25mL
Product Code. 30484450
Brand: Xylem 90525
Calibration oil for calibration of refractometers.
  • Calibration Oil
  • For Refractometers
TRUSTED_SUSTAINABILITY
Specifications
Type Calibration Fluid
Refractive Index 1.46666
For Use With (Equipment) Refractometer