missing translation for 'onlineSavingsMsg'
Learn More

Calibration Solution, Hanna Instruments™

Product Code. 10177040
Change view
Click to view available options
pH:
4.01
7.01
Unit Size:
460mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. pH Unit Size
10177040 7.01 460mL
10228700 4.01 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10177040 Supplier Hanna Instruments Supplier No. HI7007L
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

For use in calibration. Hanna Instruments™ Calibration Solution are specially formulated to have expiration of 5 years from manufacture for unopened bottle.

  • Premium quality
  • Lot number and expiration date clearly marked on label
  • Air tight with tamper-proof seal to ensure quality of solution
  • NIST traceable and supplied with Certificate of Analysis
  • Easy to identify different buffer values
  • Solution does not contain any dyes that may stain reference cell of pH electrode

Specifications

Color Undesignated
Certificate of Analysis Yes
pH 7.01
Quantity 500 mL
Traceability to NIST Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.