missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hanna Instruments™ Calibration Solution
Click to view available options
pH Range:
4.01
Unit Size:
460mL
Specifications
Specifications
| Accuracy (pH) | ±0.01 pH |
| Type | Calibration Solution |
| For Use With (Application) | Easy to identify different buffer values |
| pH Range | 4.01 |
| Certifications/Compliance | ISO 3696/BS3978 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction