missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calicivirus Antibody (FCV1-43), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-63286DL594
This item is not returnable.
View return policy
Description
Calicivirus Monoclonal antibody specifically detects Calicivirus in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Calicivirus | |
| Monoclonal | |
| DyLight 594 | |
| FCV | |
| Calcivirus viral capsid protein (63 kDa) | |
| 0.125 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG2a |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| FCV1-43 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction