Learn More
MedChemExpress Calmagite
Product Code. 16418906
Encompass
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. 16418906
Brand: MedChemExpress HYD0801
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
MedChemExpress Calmagite is a complexometric indicator which can be used to detect calcium and magnesium in various samples.
Specifications
Content And Storage 4°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light)
Product Type Dye Reagents
Quantity 100 mg
Shipping Condition Room temperature
Solubility DMSO : 33.33 mg/mL (93.00 mM; Need ultrasonic) ∣ H2O : 7.14 mg/mL (19.92 mM; Need ultrasonic)
Target HBV
Molecular Weight (g/mol) 358.37 g/mol
CAS 3147-14-6