missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calmegin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12629-1-AP-150UL
This item is not returnable.
View return policy
Description
Calmegin is a testis-specific endoplasmic reticulum chaperone protein. CLGN may play a role in spermatogeneisis and infertility.Calmegin is a testis-specific endoplasmic reticulum chaperone protein. CLGN may play a role in spermatogeneisis and infertility.Specifications
| Calmegin | |
| Polyclonal | |
| Unconjugated | |
| CLGN | |
| calmegin, CLGN | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 1047, 12745, 685504 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O14967, P52194 | |
| CLGN | |
| Calmegin Fusion Protein Ag3322 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction