missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CALML3 Antibody (004), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-90114CL1
This item is not returnable.
View return policy
Description
CALML3 Monoclonal antibody specifically detects CALML3 in Human samples. It is validated for ELISA
Specifications
| CALML3 | |
| Monoclonal | |
| CoraFluor 1 | |
| calmodulin-like 3, calmodulin-like protein 3, CaM-like protein, CLPCalmodulin-related protein NB-1 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human CALML3 (Uniprot#: P27482; Met1-Lys149). | |
| 0.1 mL | |
| Signal Transduction | |
| 810 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| ELISA | |
| 004 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction