missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CALML3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-00000-100ul
This item is not returnable.
View return policy
Description
CALML3 Polyclonal antibody specifically detects CALML3 in Human samples. It is validated for ELISA
Specifications
| CALML3 | |
| Polyclonal | |
| ELISA 1:1000-1:2000 | |
| calmodulin-like 3, calmodulin-like protein 3, CaM-like protein, CLPCalmodulin-related protein NB-1 | |
| Produced in rabbits immunized with purified, recombinant Human CALML3 (Uniprot#: P27482; Met1-Lys149) | |
| 100 μg | |
| Signal Transduction | |
| 810 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction