missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CALML3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA530232
This item is not returnable.
View return policy
Description
Recommended positive controls: A431, mouse liver. Predicted reactivity: Mouse (89%), Rat (91%), Chicken (81%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
May function as a specific light chain of unconventional myosin-10 (MYO10), also enhances MYO10 translation, possibly by acting as a chaperone for the emerging MYO10 heavy chain protein. May compete with calmodulin by binding, with different affinities, to cellular substrates. [UniProt].
Specifications
| CALML3 | |
| Polyclonal | |
| Unconjugated | |
| CALML3 | |
| 2310068O22Rik; AI326174; CALML3; calmodulin like 3; calmodulin-like 3; calmodulin-like protein 3; Calmodulin-related protein NB-1; caM-like protein; CLP | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 70405, 810 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| P27482, Q9D6P8 | |
| CALML3 | |
| Recombinant protein encompassing a sequence within the center region of human CALML3. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction