missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calmodulin 2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-14871
This item is not returnable.
View return policy
Description
Calmodulin 2 Polyclonal specifically detects Calmodulin 2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| Calmodulin 2 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| CALM2 | |
| Recombinant protein encompassing a sequence within the center region of human Calmodulin 2. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse. Expected to cross react based on sequence identity: Rat (100%), Xenopus laevis (100%), Xenopus tropicalis (100%), Rabbit (100%), Sheep (100%), Chimpanzee (100%), Bovine (100%), Guinea pig (100%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 1mg/mL | |
| Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000 | |
| CALM, CALM1, CALM3, CALML2, calmodulin, calmodulin 2 (phosphorylase kinase, delta), CAM, CAM1, CAM2, CAM3, CAMB, CAMC, CAMII, CAMIII, FLJ99410, LP7057 protein, PHKD, PHKD2, phosphorylase kinase delta | |
| Rabbit | |
| 17 kDa | |
| 0.1 mL | |
| Cancer | |
| 805 | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction