missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Calmodulin Polyclonal Antibody, Invitrogen™
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5122172

 View more versions of this product

Product Code. 17703574

  • € 453.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Calmodulin mediates the control of a large number of enzymes, ion channels, aquaporins and other proteins by Ca2+. Among the enzymes to be stimulated by the calmodulin-Ca2+ complex are a number of protein kinases and phosphatases. Together with CCP110 and centrin, is involved in a genetic pathway that regulates the centrosome cycle and progression through cytokinesis. Calmodulin is a small, highly conserved calcium binding protein found in all eukaryotic cells. With the capacity to bind up to four calcium ions, this 17 kDa protein acts as an important intracellular receptor for regulatory calcium signals. As it binds calcium, calmodulin undergoes conformational changes which can increase its affinity for target proteins. It acts both directly, through interaction with key target enzymes, and indirectly, via specific kinases. Studies have found that calmodulin participates in the regulation of several biological processes including energy and biosynthetic metabolism, cell motility, exocytosis, cytoskeletal assembly, and intracellular modulation of both cAMP and calcium concentrations. Calmodulin is the archetype of the family of calcium-modulated proteins of which nearly 20 members have been found. They are identified by their occurrence in the cytosol or on membranes facing the cytosol and by a high affinity for calcium. Calmodulin contains 149 amino acids and has 4 calcium-binding domains. Its functions include roles in growth and the cell cycle as well as in signal transduction and the synthesis and release of neurotransmitters.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Calmodulin
Polyclonal
Unconjugated
Calm1
1500001E21Rik; 2310037J09Rik; AI256814; AI327027; AI461935; AI649108; AL024000; AL024017; AY314008; calcium binding gene, DD112; calcium-binding protein Dd112; CALM; CALM 1; CALM1; CALM2; Calm3; Calm4; Calm5; CALML2; Calmodulin; calmodulin 1; calmodulin 1 (phosphorylase kinase, delta); calmodulin 2; calmodulin 2 (phosphorylase kinase, delta); calmodulin 3; calmodulin 3 (phosphorylase kinase, delta); calmodulin 4; calmodulin 5; Calmodulin III; calmodulin-1; Calmodulin-2; calmodulin-3; Calmodulin-4; calmodulin-like; calmodulin-like protein; calmodulin-like protein (neoCaM); caM; CAM1; Cam2; CAM3; CaMA; Camb; CamC; CaMI; CAMII; CAMIII; CPVT4; DD112; DD132; DD312; epididymis secretory protein Li 72; HEL-S-72; hypothetical protein; LP7057 protein; LQT14; LQT15; MGC128137 protein; neo-calmodulin; NeoCaM; PHKD; PHKD2; PHKD3; phosphorylase kinase delta; phosphorylase kinase subunit delta; phosphorylase kinase, delta subunit; prepro-calmodulin 1; prepro-calmodulin 2; prepro-calmodulin 3; R75142; RNCAMIII; Scarf; Scarf2; similar to calmodulin; skin calmodulin-related factor
Rabbit
Antigen affinity chromatography, Protein A
RUO
801, 805, 808
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.5 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
P0DP23, P0DP24, P0DP25
Calm1, Calm2, CALM3
Met1-Lys149 (Accession P62158), with N-terminal His and GST Tag.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.