missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calnexin Antibody (1C2.2D11), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-36570JF549
This item is not returnable.
View return policy
Description
Calnexin Monoclonal antibody specifically detects Calnexin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Flow (Intracellular)
Specifications
| Calnexin | |
| Monoclonal | |
| Janelia Fluor 549 | |
| calnexin, CNX, IP90FLJ26570, Major histocompatibility complex class I antigen-binding protein p88, P90 | |
| Partial recombinant human Calnexin protein (between amino acids 1-300). [UniProt P27824] | |
| 0.1 mL | |
| ER Markers, Neuroscience, Sensory Systems, Unfolded Protein Response, Vision | |
| 821 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Flow Cytometry (Intracellular Staining) | |
| 1C2.2D11 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction