missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calnexin Antibody, Janelia Fluor™ 549, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-1965JF549
This item is not returnable.
View return policy
Description
Calnexin Polyclonal antibody specifically detects Calnexin in Human, Mouse, Rat, Porcine, Avian, Bovine, Chicken, Drosophila, Guinea Pig, Rabbit, Sheep, Xenopus, Zebrafish samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| Calnexin | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Pig, Avian | |
| Purified |
| Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| Janelia Fluor 549 | |
| calnexin, CNX, IP90FLJ26570, Major histocompatibility complex class I antigen-binding protein p88, P90 | |
| A synthetic peptide made to an internal region of the canine Calnexin protein (within residues 25-100). [Swiss-Prot P24643] | |
| 0.1 mL | |
| ER Markers, Neuroscience, Sensory Systems, Unfolded Protein Response, Vision | |
| 821 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction