Learn More
Invitrogen™ Calnexin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534804
Description
Recommended positive controls: 293T, A431, HeLa, HepG2, mouse liver, PC-12, Rat2. Predicted reactivity: Mouse (97%), Rat (98%), Zebrafish (82%), Xenopus laevis (86%), Dog (98%), Pig (96%), Chicken (92%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Calnexin, also referred to as IP90, p88 and p90, is an approximately 90 kDa integral membrane protein of the endoplasmic reticulum (ER). Many resident ER proteins act as molecular chaperones and participate in the proper folding of polypeptides and their assembly into multisubunit proteins. Studies indicate that calnexin associates with the major histocompatibility complex (MHC) class I heavy chains, partial complexes of the T cell receptor and B cell membrane immunoglobulin, but not with completed receptor complexes. It has been shown that calnexin is a chaperone that retains incompletely or improperly folded proteins in the ER. The sequence Lys-Asp-Glu-Leu (KDEL) or a closely related sequence, is present at the carboxy-terminus of soluble ER resident proteins such as GRP78 and GRP94 and protein disulfide isomerase. Integral membrane ER resident proteins, like calnexin, often lack this KDEL sequence but contain positively charged cytosolic residues that ensure ER retention. Calnexin contains a large ER luminal domain (461 amino acids), a transmembrane segment (22 amino acids), and a cytoplasmic tail (89 amino acids).
Specifications
| Calnexin | |
| Polyclonal | |
| Unconjugated | |
| CANX | |
| 1110069N15Rik; AI988026; calmegin; calnexin; canx; canx-a; canx-b; clgn; clnx; cnx; D11Ertd153e; FLJ26570; hypothetical protein; IP90; Major histocompatibility complex class I antigen-binding protein p88; p88; P90; PP90; pp90 precursor | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 12330, 29144, 821 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.53 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| P27824, P35564, P35565 | |
| CANX | |
| Recombinant protein encompassing a sequence within the center region of human Calnexin. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.