Learn More
Calnexin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10427-2-AP-20UL
 View more versions of this product
Product Code. 16826813
  • € 145.00 / 20µL
Description
Calnexin, also referred to as IP90, p88 and p90, is an approximately 90 kDa integral membrane protein of the endoplasmic reticulum (ER). Many resident ER proteins act as molecular chaperones and participate in the proper folding of polypeptides and their assembly into multisubunit proteins. Studies indicate that calnexin associates with the major histocompatibility complex (MHC) class I heavy chains, partial complexes of the T cell receptor and B cell membrane immunoglobulin, but not with completed receptor complexes. It has been shown that calnexin is a chaperone that retains incompletely or improperly folded proteins in the ER. The sequence Lys-Asp-Glu-Leu (KDEL) or a closely related sequence, is present at the carboxy-terminus of soluble ER resident proteins such as GRP78 and GRP94 and protein disulfide isomerase. Integral membrane ER resident proteins, like calnexin, often lack this KDEL sequence but contain positively charged cytosolic residues that ensure ER retention. Calnexin contains a large ER luminal domain (461 amino acids), a transmembrane segment (22 amino acids), and a cytoplasmic tail (89 amino acids).
Specifications
Specifications
Calnexin
Polyclonal
Unconjugated
CANX
calnexin, CANX
Rabbit
Antigen Affinity Chromatography
RUO
12330, 29144, 821
-20°C
Liquid
Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry
0.39 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P27824, P35564, P35565
Canx
Calnexin Fusion Protein Ag0697
20 μL
Primary
Rat, Human, Mouse