missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calpain 1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | Calpain 1 |
|---|---|
| Concentration | 0.35 mg/mL |
| Applications | Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunoprecipitation, Flow Cytometry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16856983
|
Proteintech
10538-1-AP-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16846983
|
Proteintech
10538-1-AP-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The calpains including Calpain 1 (CAPN1), calcium-activated neutral proteases, are nonlysosomal, intracellular cysteine proteases. The mammalian calpains include ubiquitous, stomach-specific, and muscle-specific proteins. The ubiquitous enzymes consist of heterodimers with distinct large, catalytic subunits associated with a common small, regulatory subunit. This gene encodes the large subunit of the ubiquitous enzyme, calpain 1. Several transcript variants encoding two different isoforms have been found for this gene. Diseases associated with CAPN1 include Spastic Paraplegia 76, Autosomal Recessive and Spasticity.Specifications
| Calpain 1 | |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunoprecipitation, Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| Rat, Mouse, Human | |
| O35350, P07384, P97571 | |
| 12333, 29153, 823 | |
| Calpain 1 Fusion Protein Ag0810 | |
| Primary | |
| -20°C | |
| CAPN1 |
| 0.35 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Calpain 1, Calpain 1 catalytic subunit, Calpain 1 large subunit, Calpain mu type, CANP, CANP 1, CANP1, CANPL1, CAPN1, Micromolar calpain, muCANP, muCL | |
| Capn1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title