missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Calpain 11 Polyclonal Antibody
GREENER_CHOICE

Product Code. 13464927
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13464927 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13464927 Supplier Invitrogen™ Supplier No. PA530432

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Recommended positive controls: NT2D1, HeLa. Predicted reactivity: Mouse (81%), Chimpanzee (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The calpains including Calpain 1 (CAPN1), calcium-activated neutral proteases, are nonlysosomal, intracellular cysteine proteases. The mammalian calpains include ubiquitous, stomach-specific, and muscle-specific proteins. The ubiquitous enzymes consist of heterodimers with distinct large, catalytic subunits associated with a common small, regulatory subunit. This gene encodes the large subunit of the ubiquitous enzyme, calpain 1. Several transcript variants encoding two different isoforms have been found for this gene. Diseases associated with CAPN1 include Spastic Paraplegia 76, Autosomal Recessive and Spasticity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calpain 11
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene CAPN11
Gene Accession No. Q9UMQ6
Gene Alias calcium-activated neutral proteinase 11; calcium-activated neutral proteinase 11 {ECO:0000250; calcium-dependent thiol protease; calpain 11; calpain 11 {ECO:0000312; calpain11; Calpain-11; calpain-11; LOW QUALITY PROTEIN: calpain-11; CANP 11; CANP 11 {ECO:0000250; Capn11; capn11 {ECO:0000312; EMBL:AAH97256.1}; HIP2; LIG; RGD:1302946}; UniProtKB:Q9UMQ6}
Gene Symbols CAPN11
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Calpain 11. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11131
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.