missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calpain 6 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-99462CL1
This item is not returnable.
View return policy
Description
Calpain 6 Polyclonal antibody specifically detects Calpain 6 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Calpain 6 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| CoraFluor 1 | |
| calpain 6, calpain-6, Calpain-like protease X-linked, Calpamodulin, CalpMCALPM, CANPXcalpamodulin, CAPNX, DJ914P14.1, EC 3.4.22 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Calpain 6. | |
| 0.1 mL | |
| Cell Biology, Neuroscience, Signal Transduction | |
| 827 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction