missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Calpain 7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29697
This item is not returnable.
View return policy
Description
Calpain 7 Polyclonal antibody specifically detects Calpain 7 in Human samples. It is validated for Western Blot
Specifications
| Calpain 7 | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| calpain 7, calpain like protease, CALPAIN7, EC 3.4.22, EC 3.4.22.-, FLJ36423, homolog of Aspergillus Nidulans PALB, PalB homolog, PalBH, PALBHcalpain-7 | |
| between 50 and 100 | |
| 100 μg | |
| Cell Biology, Neuroscience, Signal Transduction | |
| 23473 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction