missing translation for 'onlineSavingsMsg'
Learn More

Calpain 9 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

€ 415.00 - € 572.00

Specifications

Antigen Calpain 9
Dilution Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18668927
View Documents
Novus Biologicals
NBP2-48545-25ul
25 μL
€ 415.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18689078
View Documents
Novus Biologicals
NBP2-48545
0.1 mL
€ 572.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Calpain 9 Polyclonal antibody specifically detects Calpain 9 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

Calpain 9
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Cell Biology, Neuroscience, Signal Transduction
PBS (pH 7.2), 40% Glycerol
10753
IgG
Immunogen affinity purified
Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000
Polyclonal
Purified
RUO
Human
calpain 9, calpain 9 (nCL-4), calpain-9, Digestive tract-specific calpain, EC 3.4.22, EC 3.4.22.54, GC36, NCL4, nCL-4EC 3.4.22.-, New calpain 4, novel calpain large subunit-4, Protein CG36
This antibody was developed against a recombinant protein corresponding to amino acids: PGEYILIPSTFEPHQEADFCLRIFSEKKAITRDMDGNVDIDLPEPPKPTPPDQETEEEQRFRALFEQVAGEDMEVTAEELEYVLNAVLQKKKDIKFKKLS
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.