missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calpain 9 Antibody (OTI3G10), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-01305
This item is not returnable.
View return policy
Description
Calpain 9 Monoclonal specifically detects Calpain 9 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Calpain 9 | |
| Monoclonal | |
| 0.8 mg/mL | |
| Western Blot 1:500-2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:150 | |
| CAPN9 | |
| Full length human recombinant protein of human CAPN9(NP_006606) produced in HEK293T cell. | |
| Affinity Purified | |
| RUO | |
| 10753 | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| OTI3G10 | |
| Unconjugated | |
| calpain 9, calpain 9 (nCL-4), calpain-9, Digestive tract-specific calpain, EC 3.4.22, EC 3.4.22.54, GC36, NCL4, nCL-4EC 3.4.22.-, New calpain 4, novel calpain large subunit-4, Protein CG36 | |
| Mouse | |
| 78.9 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction