missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calpain S1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-15677
This item is not returnable.
View return policy
Description
Calpain small subunit 1 Polyclonal specifically detects Calpain small subunit 1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Calpain small subunit 1 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| Calcium-activated neutral proteinase small subunit, Calcium-dependent protease small subunit, Calcium-dependent protease small subunit 1, calcium-dependent protease, small subunit, calpain 4, small subunit (30K), Calpain regulatory subunit, calpain small subunit 1, calpain, small polypeptide, calpain, small subunit 1, CANP, CANPS, CAPN4CALPAIN4,30K, CAPNS, CDPSCANP small subunit, CSS1 | |
| Rabbit | |
| 28 kDa | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 826 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| P04632 | |
| CAPNS1 | |
| Recombinant protein encompassing a sequence within the center region of human Calpain S1. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction