missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Calpain S1 Recombinant Rabbit Monoclonal Antibody (23GB3720), Invitrogen™

Description
Calpains are a ubiquitous, well-conserved family of calcium-dependent, cysteine proteases. Calpain families have been implicated in neurodegenerative processes, as their activation can be triggered by calcium influx and oxidative stress. Calpain I and II are heterodimeric with distinct large subunits associated with common small subunits, all of which are encoded by different genes. Calpain S1 is a small subunit common to both calpain I and II and is associated with myotonic dystrophy.
Specifications
Specifications
| Antigen | Calpain S1 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | 23GB3720 |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | CAPNS1 |
| Gene Accession No. | P04632 |
| Gene Alias | 30K; Calcium-activated neutral proteinase small subunit; calcium-dependent protease; Calcium-dependent protease small subunit; Calcium-dependent protease small subunit 1; calcium-dependent protease, small subunit; calpain 4; calpain 4, small subunit (30K); calpain II regulatory subunit; calpain II regulatory subunit (EC 3.4.22.17); calpain regulatory subunit; Calpain small subunit 1; calpain, small polypeptide; calpain, small subunit 1; CALPAIN4; CANP; CANP small subunit; CANPS; Capa4; Capa-4; Capn4; CAPNS; CAPNS1; CDPS; CSS1; D7Ertd146e; small pol; unknown protein |
| Gene Symbols | CAPNS1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?