missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calponin 1 Antibody (rCNN1/832), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54381B
This item is not returnable.
View return policy
Description
Calponin 1 Monoclonal antibody specifically detects Calponin 1 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF
Specifications
| Calponin 1 | |
| Monoclonal | |
| Biotin | |
| Basic calponin, calponin 1, basic, smooth muscle, Calponin H1, smooth muscle, calponins, basic, Sm-Calp, SMCCcalponin-1 | |
| Recombinant full-length human Calponin 1 protein (Uniprot: P51911) | |
| 100 μg | |
| Cancer, Cell Biology, Cellular Markers | |
| 1264 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF | |
| rCNN1/832 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction