missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calprotectin Mouse anti-Human, Clone: CPT/1028, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB14435.100ug
This item is not returnable.
View return policy
Description
Mouse monoclonal antibody raised against human calprotectin.
- High specificity
- High reactivity
Specifications
| Calprotectin | |
| Monoclonal | |
| Unconjugated | |
| Flow Cytometry (0.5-1 ug/million cells in 0.1 mL) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| P05109; P06702 | |
| S100A8 | |
| Recombinant protein corresponding to human calprotectin. | |
| 100 μg | |
| Primary | |
| Recognizes the L1 or calprotectin molecule, an intra-cytoplasmic antigen comprising of a 12kDa alpha chain and a 14kDa beta chain. This antibody reacts with neutrophils, monocytes, macrophages, and squamous mucosal epithelia and is important for identifying macrophages in tissue sections. | |
| Store at 4°C. | |
| IgM κ |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| CPT/1028 | |
| Mouse monoclonal antibody raised against human calprotectin. | |
| In 1 mM PBS (0.05% BSA and 0.05% azide). | |
| 60B8AG/CAGA/CFAG/CGLA/CP-10/L1Ag/MA387/MIF/MRP8/NIF/P8 | |
| Mouse | |
| Polyethylene Glycol (PEG) precipitation | |
| RUO | |
| 6279/6280 | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction