missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calreticulin-2/CALR3 Antibody (321007), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB2927MFV500
This item is not returnable.
View return policy
Description
Calreticulin-2/CALR3 Monoclonal antibody specifically detects Calreticulin-2/CALR3 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| Calreticulin-2/CALR3 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 321007 | |
| Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| calreticulin 2, calreticulin 3, Calreticulin-2, calreticulin-3, cancer/testis antigen 93, CRT2calreticulin-2, CT93, FLJ25355, MGC26577 | |
| E. coli-derived recombinant human Calreticulin-2, Thr20-Leu384, Accession # Q96L12 | |
| 0.1 mL | |
| Cancer | |
| 125972 | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction