missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calreticulin-2/CALR3 Antibody (OTI1E1), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72380AF405
This item is not returnable.
View return policy
Description
Calreticulin-2/CALR3 Monoclonal specifically detects Calreticulin-2/CALR3 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot.
Specifications
| Calreticulin-2/CALR3 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CALR3 | |
| Full length recombinant protein of human Calreticulin-2/CALR3 (NP_659483) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| IgG1 |
| Western Blot | |
| OTI1E1 | |
| Western Blot | |
| calreticulin 2, calreticulin 3, Calreticulin-2, calreticulin-3, cancer/testis antigen 93, CRT2calreticulin-2, CT93, FLJ25355, MGC26577 | |
| Mouse | |
| Affinity Purified | |
| Cancer | |
| 125972.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction