missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calreticulin Antibody (SAIC-16D-6B9), FITC , Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20094F
This item is not returnable.
View return policy
Description
Calreticulin Monoclonal antibody specifically detects Calreticulin in Human samples. It is validated for ELISA
Specifications
| Calreticulin | |
| Monoclonal | |
| FITC | |
| autoantigen Ro, CALR, calregulin, calreticulin, cC1qR, CRP55, CRT, Endoplasmic reticulum resident protein 60, epididymis secretory sperm binding protein Li 99n, ERp60, FLJ26680, grp60, HACBP, HEL-S-99n, RO, Sicca syndrome antigen A (autoantigen Ro; calreticulin), SSA | |
| Peptide "e;GLQTSQDAR"e;, derived from the calcium-binding protein chaperone Calreticulin, conjugated to KLH. | |
| 0.1 mL | |
| Autophagy, Cancer, Cellular Markers, ER Markers, Immunology, Membrane Trafficking and Chaperones, Neurodegeneration, Neuroscience, Signal Transduction | |
| 811 | |
| Store at 4°C in the dark. | |
| IgG κ |
| ELISA | |
| SAIC-16D-6B9 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction