Learn More
Calreticulin (CALR) Monoclonal Antibody (OTI13E4), OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA813228S
Encompass_Preferred
Product Code. 17508004
  • € 242.00 / 30µL
Description
Calreticulin (CALR) Monoclonal antibody specifically detects Calreticulin (CALR) in Human samples. It is validated for Western Blot
Specifications
Specifications
Calreticulin (CALR)
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P19838
Mouse
Affinity chromatography
RUO
4790
-20°C
IgG1
Western Blot
OTI13E4
Unconjugated
NFKB1
Autoantigen RO; CABP3; CALBP; calcium-binding protein 3; CALR; calrectulin; calregulin; Calreticulin; calreticulin precursor; cC1qR; CRP55; CRT; CRTC; Endoplasmic reticulum resident protein 60; epididymis secretory sperm binding protein Li 99n; ERp60; FLJ26680; grp60; HACBP; HEL-S-99n; I79_008346; RO; Ro/SS-A Antigen; Sicca syndrome antigen A (autoantigen Ro; calreticulin); SSA
Full length human recombinant protein of human NFKB1 (NP_003989) produced in HEK293T cell.
30 μL