Learn More
Calreticulin (CALR) Monoclonal Antibody (OTI13E4), OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA813228S
This item is not returnable.
View return policy
Description
Calreticulin (CALR) Monoclonal antibody specifically detects Calreticulin (CALR) in Human samples. It is validated for Western BlotSpecifications
| Calreticulin (CALR) | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P19838 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 4790 | |
| -20°C | |
| IgG1 |
| Western Blot | |
| OTI13E4 | |
| Unconjugated | |
| NFKB1 | |
| Autoantigen RO; CABP3; CALBP; calcium-binding protein 3; CALR; calrectulin; calregulin; Calreticulin; calreticulin precursor; cC1qR; CRP55; CRT; CRTC; Endoplasmic reticulum resident protein 60; epididymis secretory sperm binding protein Li 99n; ERp60; FLJ26680; grp60; HACBP; HEL-S-99n; I79_008346; RO; Ro/SS-A Antigen; Sicca syndrome antigen A (autoantigen Ro; calreticulin); SSA | |
| Full length human recombinant protein of human NFKB1 (NP_003989) produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |