missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Calreticulin Monoclonal Antibody (1G6A7)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA515382

Product Code. 17707597

  • € 568.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MA5-15382 targets Calreticulin in IF, IHC, Flow and WB applications and shows reactivity with Human, non-Human primate, and mouse samples. The MA5-15382 immunogen is synthetic peptide corresponding to aa (EEEDVPGQAKDELC) of human Calreticulin, conjugated to KLH. MA5-15382 detects Calreticulin which has a predicted molecular weight of approximately 48kDa.

Calr or Calreticulin is a calcium-binding chaperone that promotes folding, oligomeric assembly and quality control in the endoplasmic reticulum (ER) via the calreticulin/calnexin cycle. Calr, a lectin, interacts transiently with almost all of the monoglucosylated glycoproteins that are synthesized in the ER. Calr also interacts with with the DNA-binding domain of NR3C1 and mediates its nuclear export. It may also be involved in maternal gene expression regulation and oocyte maturation via the regulation of calcium homeostasis.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Calreticulin
Monoclonal
1 mg/mL
PBS with 0.05% sodium azide
P14211, P27797
CALR
Synthetic peptide corresponding to aa (EEEDVPGQAKDELC) of human Calreticulin, conjugated to KLH.
100 μg
Primary
Human, Mouse, Monkey
Antibody
IgG2a
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1G6A7
Unconjugated
CALR
Autoantigen RO; CABP3; CALBP; calcium-binding protein 3; CALR; calrectulin; calregulin; Calreticulin; calreticulin precursor; cC1qR; CRP55; CRT; CRTC; Endoplasmic reticulum resident protein 60; epididymis secretory sperm binding protein Li 99n; ERp60; FLJ26680; grp60; HACBP; HEL-S-99n; I79_008346; RO; Ro/SS-A Antigen; Sicca syndrome antigen A (autoantigen Ro; calreticulin); SSA
Mouse
Protein A
RUO
12317, 811
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.