Learn More
Invitrogen™ Calreticulin Recombinant Rabbit Monoclonal Antibody (23GB700), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA553203
 View more versions of this product
Product Code. 30089483
  • € 207.00 / 20µL
Description
Calr or Calreticulin is a calcium-binding chaperone that promotes folding, oligomeric assembly and quality control in the endoplasmic reticulum (ER) via the calreticulin/calnexin cycle. Calr, a lectin, interacts transiently with almost all of the monoglucosylated glycoproteins that are synthesized in the ER. Calr also interacts with with the DNA-binding domain of NR3C1 and mediates its nuclear export. It may also be involved in maternal gene expression regulation and oocyte maturation via the regulation of calcium homeostasis.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Calreticulin
Recombinant Monoclonal
0.3 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P14211, P18418, P27797
CALR
A synthetic peptide of human calreticulin (45-86AA).
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
23GB700
Unconjugated
CALR
Autoantigen RO; CABP3; CALBP; calcium-binding protein 3; CALR; calrectulin; calregulin; Calreticulin; calreticulin precursor; cC1qR; CRP55; CRT; CRTC; Endoplasmic reticulum resident protein 60; epididymis secretory sperm binding protein Li 99n; ERp60; FLJ26680; grp60; HACBP; HEL-S-99n; I79_008346; RO; Ro/SS-A Antigen; Sicca syndrome antigen A (autoantigen Ro; calreticulin); SSA
Rabbit
Affinity chromatography
RUO
12317, 64202, 811
-20°C