missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calretinin Antibody (3G9), Novus Biologicals™
Mouse Monoclonal Antibody
€ 180.00 - € 499.00
Specifications
| Antigen | Calretinin |
|---|---|
| Clone | 3G9 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000 - 1:5000, Immunohistochemistry 1:1000, Immunocytochemistry/ Immunofluorescence 1:1000, Immunohistochemistry Free-Floating |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Free Floating) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18685276
|
Novus Biologicals
NBP2-50042 |
0.1 mL |
€ 499.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18618816
|
Novus Biologicals
NBP2-50042SS |
0.025 mL |
€ 180.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beskrivning
Calretinin Monoclonal antibody specifically detects Calretinin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry Free-FloatingSpecifikationer
| Calretinin | |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Free Floating) | |
| Unconjugated | |
| Mouse | |
| Cellular Markers | |
| 50% PBS, 50% glycerol | |
| 29 kDa calbindin, CAB29, CAL2, calbindin 2, calbindin 2, (29kD, calretinin), calbindin 2, 29kDa (calretinin), calbindin D29K, calretinin, CR | |
| Full-length recombinant human Calretinin expressed in and purified from E. coli. [UniProt# P22676] | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 3G9 | |
| Western Blot 1:1000 - 1:5000, Immunohistochemistry 1:1000, Immunocytochemistry/ Immunofluorescence 1:1000, Immunohistochemistry Free-Floating | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| P22676 | |
| 794 | |
| IgG1 | |
| Immunogen affinity purified |
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel