missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calretinin Antibody (S01-2G7), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 422.00 - € 647.00
Specifications
| Antigen | Calretinin |
|---|---|
| Clone | S01-2G7 |
| Concentration | 0.3 mg/mL |
| Applications | Western Blot, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18661194
|
Novus Biologicals
NBP3-19588-100ul |
100 μg |
€ 647.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18653835
|
Novus Biologicals
NBP3-19588-50ul |
50 μg |
€ 422.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Calretinin Monoclonal antibody specifically detects Calretinin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| Calretinin | |
| 0.3 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| 29 kDa calbindin, CAB29, CAL2, calbindin 2, calbindin 2, (29kD, calretinin), calbindin 2, 29kDa (calretinin), calbindin D29K, calretinin, CR | |
| A synthetic peptide of human Calretinin (Uniprot # P22676) | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
| S01-2G7 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cellular Markers, Phospho Specific | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 794 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title