missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Calretinin Monoclonal Antibody (6A9)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA547457
This item is not returnable.
View return policy
Description
Calretinin Monoclonal Antibody for Western Blot, ICC/IF, IHC (PFA)
Calretinin is encoded by the CALB2 gene located on chromosome 16. It is a troponin C superfamily protein having six EF-hand structural motifs. The EF motif allows the protein to bind calcium, and function in calcium signaling, calcium buffering, and modulating neuronal excitability. Calretinin is expressed in the mesothelium, mast cells, some neural cells, hair follicles, and fat cells.
Specifications
| Calretinin | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and no preservative | |
| P22676, P47728, Q08331, Q3ZBY3 | |
| CALB2 | |
| Full-length recombinant human protein expressed in and purified from E. coli. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Bovine, Pig, Horse | |
| Antibody | |
| IgA |
| Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry | |
| 6A9 | |
| Unconjugated | |
| CALB2 | |
| 29 kDa calbindin; 29kD calretinin; 29kDa calbindin; CAB29; CAL2; Calb2; calbindin 2; calbindin 2, (29kD, calretinin); calbindin 2, 29kDa; calbindin D29K; calretinin; CR; testicular secretory protein Li 8 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 100068401, 100127479, 117059, 12308, 513947, 794 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction