Learn More
Calretinin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12278-1-AP-150UL
 View more versions of this product
Product Code. 16809903
  • € 410.00 / 150µL
Description
Calretinin is encoded by the CALB2 gene located on chromosome 16. It is a troponin C superfamily protein having six EF-hand structural motifs. The EF motif allows the protein to bind calcium, and function in calcium signaling, calcium buffering, and modulating neuronal excitability. Calretinin is expressed in the mesothelium, mast cells, some neural cells, hair follicles, and fat cells.
Specifications
Specifications
Calretinin
Polyclonal
Unconjugated
CALB2
29 kDa calbindin, CAB29, CAL2, CALB2, calbindin 2, Calretinin, CR
Rabbit
Antigen Affinity Chromatography
RUO
117059, 12308, 794
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.25 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P22676, P47728, Q08331
CALB2
Calretinin Fusion Protein Ag2924
150 μL
Primary
Human, Mouse, Rat