missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Calretinin Recombinant Rabbit Monoclonal Antibody (RM324)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA544580
This item is not returnable.
View return policy
Description
Calretinin Recombinant Monoclonal Antibody for IHC (P), Western Blot
Calretinin is encoded by the CALB2 gene located on chromosome 16. It is a troponin C superfamily protein having six EF-hand structural motifs. The EF motif allows the protein to bind calcium, and function in calcium signaling, calcium buffering, and modulating neuronal excitability. Calretinin is expressed in the mesothelium, mast cells, some neural cells, hair follicles, and fat cells.
Specifications
| Calretinin | |
| Recombinant Monoclonal | |
| 200 μg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.09% sodium azide | |
| P22676, P47728, Q08331 | |
| CALB2 | |
| A peptide corresponding to N-terminus of human Calretinin. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| RM324 | |
| Unconjugated | |
| CALB2 | |
| 29 kDa calbindin; 29kD calretinin; 29kDa calbindin; CAB29; CAL2; Calb2; calbindin 2; calbindin 2, (29kD, calretinin); calbindin 2, 29kDa; calbindin D29K; calretinin; CR; testicular secretory protein Li 8 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 117059, 12308, 794 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction