missing translation for 'onlineSavingsMsg'
Learn More

Calsequestrin 1 Antibody, Novus Biologicals™

Product Code. 18623188 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18623188 100 μg 100µL
18649287 50 μg 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18623188 Supplier Novus Biologicals Supplier No. NBP300194100ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Calsequestrin 1 Polyclonal antibody specifically detects Calsequestrin 1 in Human samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calsequestrin 1
Applications ELISA, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution ELISA 1:5000-1:10000, Immunocytochemistry/ Immunofluorescence 1:100-1:500, Immunohistochemistry-Paraffin 1:200-1:1000
Formulation 0.2 um filtered solution in PBS
Gene Alias calmitin, calmitine, calsequestrin 1 (fast-twitch, skeletal muscle), Calsequestrin, skeletal muscle isoform, calsequestrin-1, CASQ, PDIB1
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human Calsequestrin 1 (Accession#: P31415; Gln35-Asp396)
Purification Method Antigen and protein A Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 844
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.