Learn More
Calumenin Mouse anti-Human, Unconjugated, Clone: 2D8B8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67585-1-IG-20UL
Description
The Calumenin antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Calumenin. This antibody recognizes human, mouse, rat antigen. The Calumenin antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
| Calumenin | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Western Blot, Immunofluorescence, ELISA | |
| 2D8B8 | |
| Unconjugated | |
| 20 μL | |
| 813 | |
| IgG1 |